Learn More
Surgical Design™ Frazier Dissecting Hook
Catalog No. 21124584 Shop All Surgical Design Products
Change view
Click to view available options
Style:
Hook
Micro Hook
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
21-124-584 Hook
21-124-429 Micro Hook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-124-584 Supplier Surgical Design™ Supplier No. 2904M
Add to Cart
Add to Cart
Frazier dissecting hook delivers fine precision for soft tissue manipulation and nerve isolation in delicate and confined applications.
  • Made from high-quality surgical-grade stainless steel for superior strength, balance, and corrosion resistance
  • Available in Standard and Micro hook sizes to accommodate various surgical applications
  • 5-in. overall length ensures controlled reach and precision during fine dissection
  • Delicately curved hook tip allows for safe tissue retraction and nerve exposure without trauma
  • Ergonomic, ribbed handle provides excellent grip and maneuverability
  • Polished surface finish for reduced glare and smooth tissue interaction
  • Lightweight and balanced design minimizes hand fatigue during prolonged procedures
  • Autoclavable and reusable, ensuring consistent reliability across multiple sterilization cycles
  • Crafted for high precision in micro and standard surgical dissection
  • Ideal for microsurgery, ENT, and neuro procedures
  • Commonly used for tissue separation, nerve isolation, and fine dissection
  • Suitable for medical, anatomical, and laboratory settings
Specifications
Product Type Micro Hook
Autoclavable Autoclavable
Color Silver
Material Stainless Steel
Style Hook
Length (English) 5 in.
Length (Metric) 12.7 cm
Sterility Non-sterile