Learn More
Med Vet International FRCPSSEMKENDRESSING5SERR
Supplier:  Med Vet International MILMH6104
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Semken Dressing Forceps. Semken Dressing (Thumb) Forceps are used for holding dressing materials such as cotton and gauze during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 5in, Serrated, MH6-104
Catalog No. 50-255-9499
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.