Learn More
Med Vet International FRCPSTISSSERRHDLE4.5GER
Supplier:  Med Vet International MIL640
Encompass_Preferred
The Miltex 6-40 tissue forceps are made of OR grade German stainless steel and measure 4.5" in length. They have a non-locking, serrated thumb handles and the tips are straight with 1 x 2 teeth.
Catalog No. 50-255-9539
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.