Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Free Fatty Acid Uptake Assay Kit, Fluorometric, 100 Tests

Catalog No. 502582275
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2275 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2275 Supplier Abcam Supplier No. AB176768100TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorometric assay for quantifying free fatty acid uptake in adherent cells within one hour.

  • Assay Type: Cell-based
  • Detection Method: Fluorescent
  • Sample Types: Adherent cells
  • Assay Time: 1 hr
  • Results Type: Quantitative
  • Performance: Cited in > 22 publications
  • Compatible Equipment: Microplate reader

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Free Fatty Acids
Detection Location In-Cell Detection
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 100 Tests
Run Time 1 hr.
Substrate Properties Lipid-based Substrate
Detection Method Fluorescence
Sample Type Adherent cells
Quantity 100 Tests
For Use With (Application) Free fatty acid uptake assay
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.