Learn More
Crystal Chem Inc Free Prostate-Specific Antigen (fPSA) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80977
Encompass
The Free Prostate-Specific Antigen (fPSA) ELISA is a highly sensitive assay that is used to quantify levels of human free PSA in serum. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 0.05 ng/mL. Dynamic range of 0.1 - 15 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.