Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Free T3 (Triiodothyronine) ELISA Kit

Catalog No. MP07BC1006 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
96 Wells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07BC1006 96 Wells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP07BC1006 Supplier MP Biomedical™ Supplier No. 07BC1006

Quantitative Enzyme-linked Immunosorbent Assay

The Free T3 (Triiodothyronine) ELISA Kit is intended for the measurement of Free T3 in a sample.
TRUSTED_SUSTAINABILITY

Specifications

Description Intended for the measurement of Free T3 in a sample.
Product Type Kit and Assay
Sample Type Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 96 Wells
Regulatory Status FDA Cleared
Sample Volume 50 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.