Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Freeze Protected, Wall Mounted Drench Shower
GREENER_CHOICE

Catalog No. 18666687 Shop All Justrite Safety Group Products
Change view
Click to view available options
Electrical Requirements:
120 V
240 V
Material (Piping Connection):
Galvanized Steel
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements Material (Piping Connection)
18-666-687 120 V Galvanized Steel
18-666-688 120 V Stainless Steel
18-666-689 240 V Galvanized Steel
18-666-690 240 V Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-666-687 Supplier Justrite Manufacturing Co Supplier No. H2G1H
Add to Cart
Add to Cart

Hughes Freeze Protected, Wall Mounted Drench Shower is intended for use where there is a possibility of the water inside of the shower freezing due to extreme cold climates

Features:

  • Trace tape heating with polyethylene jacket and polyurethane foam insulation to improve frost protection and strength
  • Ideal for use where space is at a premium
  • Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel to endure the harshest environments and site conditions
  • Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur
  • Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure
  • Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
  • Includes a removable, self-draining overhead nozzle designed for high flow rates that meet or exceed ANSI standards
  • Made of durable nylon material, the nozzle offers optimal spray distribution for user protection and water conservation.
  • For cold climates with a minimum ambient operating temperature of -4°F (-20°C) or above
  • Suitable for both Class 1 Division 2 hazardous areas, as well as non-hazardous areas
  • Includes a universal safety shower sign for enhanced visibility
  • Meets ANSI Z358.1 2014 and EN 15154 standards, supports OSHA requirements

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty and Services

1 Year

Specifications

Activation Pull Handle
Certifications/Compliance ANSI Z358.1-2014 and EN15154 Compliant
Color White with Green Stripes
Depth (English) 12 in.
Depth (Metric) 305 mm
Electrical Requirements 120 V
For Use With (Application) Emergency Shower and Eye Wash with Water
Height (English) 12 in.
Height (Metric) 305 mm
Includes 1 Unit
Inlet Connection (Metric) 1 in. (F) NPT
Material Polyurethane Foam, MDPE, Nylon 6
Material (Piping Connection) Galvanized Steel
Mounting Wall
Pressure 22 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 40 in.
Width (Metric) 1016 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.