Learn More
Eppendorf 3P FREEZER DIVIDER 7X7
Supplier:  Eppendorf 3P D100
Encompass_Preferred
Freezer box divider, 10 x 10, 11.8 mm max tube diameter, holds 100 tubes • Available in 7 × 7, 8 × 8, 9 × 9 and 10 × 10 configurations • Compatible with freezer cardboard storage boxes (sold separately)
Catalog No. 50-118-0173
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.