Learn More
Darwin Chambers Company Double Door Lab Freezer 47 cu. ft.
SDP
Supplier:  Darwin Chambers Company FZ055SS
Encompass_Preferred
The FZ055-SS is a 50 cubic foot double door laboratory freezer. Temperature range is -25°C to 15°C. This unit comes with 6 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The FZ series freezer chambers comes standard with the following performance: Solid-State Temperature Control. Autodefrost. Standard Microprocessor Controller with PID, Autotuning, programmable alarms (hi/lo), ability to ramp/soak, process variable and setpoint variable offset.
Catalog No. 50-189-3874
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.