Learn More
Thermo Scientific™ Freezer and Refrigerator Shelves
GSA_VA
Catalog No. 12370607
Change view
Click to view available options
For Use With (Equipment):
23.3, 45.8, 51.1, 78.8 cu. ft. Units
29.2 cu. ft. Units
4.7 cu. ft., 11.5 cu. ft. Units
51.1 cu. ft. Units
Type:
Open Wire Shelves
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
12-370-607 29.2 cu. ft. Units Open Wire Shelves
13-990-163 23.3, 45.8, 51.1, 78.8 cu. ft. Units Open Wire Shelves
13-990-164 51.1 cu. ft. Units Open Wire Shelves
12-370-653 4.7 cu. ft., 11.5 cu. ft. Units Open Wire Shelves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-370-607 Supplier Thermo Scientific™ Supplier No. 6695
Add to Cart
Add to Cart
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Open Wire Shelves
Exterior Finish Epoxy-coated Steel
For Use With (Equipment) 29.2 cu. ft. Units
Warranty Warranty varies by country, please contact us for details
Holds General Freezer Items