Learn More
Labconco™ FreeZone™ 18L -50°C Console Freeze Dryers w/Stoppering Tray Dryer, 230V Models



FreeZone 18L Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
$77600.00 - $78500.00
Specifications
Type | Freeze Dryer/Lyophilizer |
---|---|
Capacity (Metric) | 18 L (before defrosting) |
No. of Shelves | 3 |
Temperature Range (Metric) | -50°C |
Temperature Range (English) | -58°C |
Requires: Vacuum pump
Catalog Number | Mfr. No. | Includes | Plug Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Plug Type | Price | Quantity & Availability | ||||
10-400-394
![]() ![]() |
Labconco™
701821010 |
Stainless steel collector, Stoppering Tray Dryer | North America |
Each for $77,600.00
|
|
||||
10-400-395
![]() |
Labconco™
701821015 |
Stainless steel collector, Stoppering Tray Dryer | Saudi Arabia |
Each for $77,600.00
|
|
||||
10-400-396
![]() |
Labconco™
701821030 |
Stainless steel collector, Stoppering Tray Dryer | Schuko |
Each for $77,600.00
|
|
||||
10-400-397
![]() |
Labconco™
701821040 |
Stainless steel collector, Stoppering Tray Dryer | China/Australia |
Each for $77,600.00
|
|
||||
10-400-398
![]() |
Labconco™
701821050 |
Stainless steel collector, Stoppering Tray Dryer | British (UK) |
Each for $77,600.00
|
|
||||
10-400-399
![]() |
Labconco™
701821070 |
Stainless steel collector, Stoppering Tray Dryer | India |
Each for $77,600.00
|
|
||||
10-400-400
![]() ![]() |
Labconco™
701822010 |
PTFE-coated collector, Stoppering Tray Dryer | North America |
Each for $78,500.00
|
|
||||
10-400-401
![]() |
Labconco™
701822015 |
PTFE-coated collector, Stoppering Tray Dryer | Saudi Arabia |
Each for $78,500.00
|
|
||||
10-400-402
![]() |
Labconco™
701822030 |
PTFE-coated collector, Stoppering Tray Dryer | Schuko |
Each for $78,500.00
|
|
||||
10-400-404
![]() |
Labconco™
701822050 |
PTFE-coated collector, Stoppering Tray Dryer | British (UK) |
Each for $78,500.00
|
|
||||
10-400-405
![]() |
Labconco™
701822070 |
PTFE-coated collector, Stoppering Tray Dryer | India |
Each for $78,500.00
|
|
||||
10-400-403
![]() |
Labconco™
701822040 |
PTFE-coated collector, Stoppering Tray Dryer | China/Australia |
Each for $78,500.00
|
|
||||
Description
FreeZone 18L Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
- Upright stainless steel collector coil holds 18L of ice before defrosting
- -50° C collector is suitable for aqueous samples
- Environmentally-friendly HCFC/CFC-free refrigeration system
- Internal coil maximizes vapor-trapping efficiency
- Optional PTFE-coated collectors withstand corrosive compounds
- Lyo-Works OS provides a real time display of collector temperature and vacuum level
- 5 in. full color touch screen display
- Flexible programming and control of optional drying accessories
- Optional End-Zone™ end point detection for flask freeze drying
- One-touch data logging and display provides information to maximize sample quality and minimize run time
- Customizable settings include language, time zone, temperature and vacuum
- Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
- Front viewing window makes monitoring vacuum pump oil hassle free
- FreeZone exclusive features protect your vacuum pump from costly damage
- Vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run
- Features hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection
- Console provides interior space for a vacuum pump (sold separately)
FreeZone Stoppering Tray Dryer provides precise control and program flexibility for consistent performance.
- Three 16.5 x 12 in. temperature-controlled (±1° C), adjustable shelves with sample temperature sensors provide up to 594 sq. inches of shelf space
- Circulating fluid in shelves ranges from -40 and +40° C
- Environmentally-friendly HCFC/CFC-free refrigeration system
- Built-in pneumatic mechanism stoppers containers on all three shelves
- 1/8 in. OD backfill port introduces inert gas into the chamber, protecting samples from atmospheric moisture and contaminants
Vacuum pump with a displacement of at least 163 liters/minute and freeze dry glassware (if not bulk freeze drying) are required (not included).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
All models conform to UL Standard 61010-1 (60Hz models), CAN/CSA C22.2 No. 61010-1 (60Hz models), and CE Conformity marking (230V, 50Hz models).
Warranty and Services
One year
Specifications
Freeze Dryer/Lyophilizer | |
3 | |
-58°C | |
UL, CAN/CSA, CE | |
75.1 cm | |
82.2 cm | |
161.7 cm | |
713 lb. |
18 L (before defrosting) | |
-50°C | |
Steel | |
29.6 in. | |
32.4 in. | |
63.7 in. | |
230 V | |
323.41 kg |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.