Learn More
Essendant D'vour Clean-up Kit, Powder, All Inclusive Kit, 6/Carton
Supplier:  Essendant BGD169
Encompass_Preferred
The BIG D D-VOUR KIT transforms a liquid spill into an odorless, semi-solid mass for disposal. Effective for spills of blood, urine, feces, food and other malodor sources, this kit contains all the necessary items for clean up and disposal of spilled bodily fluids following the guidelines of OSHA and The Centers for Disease Control. Each D-VOUR KIT contains: 1 packet of D-VOUR (absorbs 80 to 100 times its weight), 1 pair of disposable gloves, 2 disposable scoops with hand grips, 1 black disposable plastic bag, 1 red biohazard disposable plastic bag, 1 germicidal disposable wipe, 2 antiseptic disinfectant towelettes, and instructions for clean up and disposal of spilled bodily fluids which follow the guidelines of OSHA and The Centers for Disease Control.
Catalog No. 27-936-92
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.