Learn More
STEMCELL Technologies Human Peripheral Blood Mononuclear Cells, Fresh, Size: 1 x 10^8 cells
SDP
Supplier:  STEMCELL Technologies 2000077
Encompass_Preferred
Primary human mononuclear cells (MNCs) were isolated from peripheral blood (PB) leukapheresis samples using density gradient separation or red blood cell lysis. Cells were obtained using Institutional Review Board (IRB)-approved consent forms and protocols.
Catalog No. 50-204-5567
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.