Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Fresnel Lens
SDPSCI_ED

Catalog No. S27803 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Vinyl Sleeve
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27803 Vinyl Sleeve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27803 Supplier United Scientific Supplies Supplier No. FRESLNS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

With a Vinyl Sleeve

  • Magnification: 1.25x
  • Magnifying Window: 3.125 x 1.75 in.
  • Overall Product Dimensions: 3.375 x 2.25 in.

Specifications

Product Type Lens, Fresnel
Includes Vinyl Sleeve
Diameter (English) Lens 3.125 x 1.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.