Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ O-Cedar™ Microfiber Flip Mop™

Catalog No. 19200629 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue, White, Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-629 Blue, White, Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-629 Supplier Vileda Professional™ Supplier No. 133950

Enjoy twice the cleaning power with this mop. Chemical-free cleaning is easy with our microfiber technology so you can clean effectively using a dry or damp mop head.

  • Eco-friendly
  • Mop head rotates 360° to clean hard to reach areas
  • Machine washable
  • Easily attracts, dust, dirt and hair from any hard surface
  • Scrubbing strips safely remove embedded dirt, which gives you twice the cleaning power
  • Perfect for quick cleans in offices, small retailers, etc

Specifications

Product Type Flip Mop
Color Blue, White, Red
For Use With (Application) Quick cleans in labs, animal research facilities, production areas
Material Microfiber Pad
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.