Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ PVAmicro Cloth

Catalog No. 19200698 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Blue
Green
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-698 Red
19-200-697 Blue
19-200-700 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-200-698 Supplier Vileda Professional™ Supplier No. 143586
Add to Cart
Add to Cart
This item is not returnable. View return policy

Clean and wipe dry in one single movement! PVAmicro™ has a high cleaning performance as well as high absorbency to wipe the surface dry without leaving streaks or droplets behind. The cloth is made from PVA for absorbency and microfiber for performance, allowing you to wipe dry and clean in one single movement.

  • Knitted microfiber ensures perfect results on almost all surfaces: glass, labs, stainless steel and more
  • The PVA as very good water absorbency, meaning streak-free wiping without after-drying
  • PVAmicro™ holds 40 times less particle residue after rinsing, compared to knitted microfiber cloth
  • Streak and scratch free performance

Specifications

Product Type PVAmicro Cloth
Color Red
Dimensions (L x W) 38 x 35 cm
For Use With (Application) For cleaning all types of hard surfaces
Packaging Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.