Learn More
Vileda Professional Roll-O-Matic Self-Wringing Mops
Suitable for all controlled environments.
Manufacturer: Vileda Professional 118534
Description
- Mop with built-in wringer
- Wringer built into handle, eliminating need for expensive and bulky wringer
- Improved components eliminate sharp edges and provide for excellent refill retention
- Fits all original and advanced technology Roll-O-Matic approved refills
- Ergonomic grip
Specifications
10 in. | |
Blue | |
48 in. | |
Galvanized Steel Mops with Sponge Mop Head, Straight Head |
Not Autoclavable | |
For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique | |
Galvanized Steel |