Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ Roll-O-Matic™ Sponge Mop Refills

Catalog No. 19200711 Shop All Vileda Professional Products
Change view
Click to view available options
Material:
CE Universal Stainless Steel
Galvanized Steel
Microfiber, Stainless Steel
Polyester with Stainless Steel Channel
Polyester, Stainless Steel
Stainless Steel
Width (English):
10 in.
14 in.
Description:
“Clean”
“Disinfect”
“Original”
“Universal”
Sterility:
Non-sterile
Sterile
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Width (English) Description Sterility
19-200-711 Stainless Steel 14 in. “Original” Sterile
19-032-1020 Stainless Steel 14 in. “Universal” Non-sterile
19-032-1021 Galvanized Steel 14 in. “Universal” Non-sterile
19-032-1022 Polyester with Stainless Steel Channel 14 in. “Disinfect” Non-sterile
19-032-1023 Microfiber, Stainless Steel 14 in. “Clean” Non-sterile
19-032-1005 Galvanized Steel 10 in. “Original” Non-sterile
19-032-1000 Galvanized Steel 14 in. “Original” Non-sterile
18-999-2406 Stainless Steel 14 in. “Original” Non-sterile
19-032-1036 Stainless Steel 10 in. “Universal” Non-sterile
19-032-1030 Stainless Steel 10 in. “Original” Non-sterile
19-200-713 CE Universal Stainless Steel 14 in. “Universal” Sterile
19-032-1037 Galvanized Steel 10 in. “Universal” Non-sterile
19-141-904 Microfiber, Stainless Steel 10 in. “Clean” Non-sterile
19-141-905 Polyester, Stainless Steel 10 in. “Disinfect” Non-sterile
19-200-712 Stainless Steel 10 in. “Universal” Sterile
19-000-669 Stainless Steel 14 in. “Disinfect” Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-200-711 Supplier Vileda Professional™ Supplier No. 147547
This item is not returnable. View return policy

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

All Vileda Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. They are ideal for general use, absorbing spills, applying disinfectant, and cleaning particulates.

  • Fits all original and self-wringing Roll-O-Matic™ mops
  • 10- or 14-inch options for both stainless steel and galvanized steel handles
  • Completely autoclavable and resistant to gamma and ETO sterilization
  • Individually packaged and lot number controlled
  • Certificate of conformance and technical data available upon request
  • Sterile versions validated to 10-6 per ISO 11137, individually double bagged with lot number and indicator dot, and supplied with certificate of sterility upon request

Certifications

Certificate of Sterility

Specifications

Color Blue
Sterility Sterile
Autoclavable Autoclavable
Width (English) 14 in.
Quantity 12/Cs.
Description “Original”
For Use With (Application) For General Purpose
Material Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.