Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ UltraSpeed Trio Mop Pad

Catalog No. 19200608 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
White, Gray, Beige
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-200-608 White, Gray, Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-608 Supplier Vileda Professional™ Supplier No. 143202

Cleans difficult areas and offers maximum cleaning performance while significantly reducing mopping times. Vileda Professional-FHP UltraSpeed Trio Mop Pad is part of the UltraSpeed Pro System and was developed to be simple to use.

  • Unique press wrings out more water than traditional gear presses, allowing floors to dry faster and be back in use quicker
  • Strip fastening of mops eliminates need to remove them for wringing
  • Looped yarn construction takes the shape of any floor, removing trapped dirt and the external fringes collect and remove all loose dirt particles
  • Excellent for spill pick up with the capacity to pick up over 500 ml in between each wringing process saving time
  • Highly durable and machine washable over 500 times at 95°C
  • Triple zone construction:
      • White microfibre zone – Microfiber for high mechanical cleaning action-minimizes chemical use
      • Gray zone – Polyester provides low friction and high durability
      • Beige zone – Blended polyester and viscose to provide absorbency

Specifications

Product Type Trio Mop Pad
Color White, Gray, Beige
Length (English) 16 in.
Quantity 20/Cs.
Description Proven 99.9% Bacteria Reduction
For Use With (Application) Surface Cleaning in LAR, Production, and Lab Environments
Material Microfiber, Polyester, Blended Polyester, Viscose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.