Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Friction Apparatus
SDPSCI_ED

Catalog No. S139501 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139501 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139501 Supplier United Scientific™ Supplier No. FRAP01
Add to Cart
Add to Cart
This item is not returnable. View return policy

Simple but complete friction demonstrator.

This set includes a friction block and a friction board, both made of high quality, smoothly finished pine wood.

  • The friction block has a hook for a spring scale and three compartments for additional weights
  • It can be placed face-up or sideways
  • Students can vary the weight or area of contact to test their impact on the frictional force
  • The set comes with three 50 g weights and a 100 g spring scale
  • The board is 20 in. x 2 in. and the block is 4 in. x 1.5 in. x 1 in.
  • Includes an activity guide

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.