Learn More
United Scientific™ Friction Box
SDPSCI_ED
Catalog No. S15779 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Product Type: Hooke's Law Apparatus
Catalog No.
S15779 Product Type: Hooke's Law Apparatus
1 options
Catalog No. S15779 Supplier United Scientific™ Supplier No. FRBX01
Add to Cart
Add to Cart
This friction box is used for coefficient of friction studies.
  • This friction box is used for coefficient of friction studies when larger loads are required.
  • Box will accommodate a load of 2 kg or more
  • Smooth sanded wood with two strips of polymer-based material attached to the bottom
  • Dimensions: 6 x 3.5 x 2.5 in.
Specifications
Product Type Hooke's Law Apparatus
For Use With (Application) Demonstration