Learn More
United Scientific™ Friction Rods and Pads
SDPSCI_ED
Catalog No. S16088 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Cotton
Glass
Plastic
Rubber
Silk
Length (English):
6 in.
12 in.
Description:
Hard Rod
Hollow Rod
Pad
Solid Rod
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Length (English) Description
S16088 Cotton 12 in. Pad
S15910 Glass 12 in. Hollow Rod
S16022 Glass 12 in. Solid Rod
S16063 Silk 12 in. Pad
S15893 Rubber 6 in. Hard Rod
S15782 Plastic 12 in. Solid Rod
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S16088 Supplier United Scientific™ Supplier No. RPCT30
Add to Cart
Add to Cart
Ideal tools for demonstrating static electricity.
  • Friction rods and pads made from various materials.
  • All the essential materials for student exploration in electrostatics
  • Topics include static electricity, polarity, and friction studies
  • Use a variety of friction rods, friction pads, and pith balls
  • The combination of materials will determine the charge distribution after contact is made
Specifications
Product Type Friction Rods and Pads
Description Pad
Material Cotton
For Use With (Application) Friction Demonstrations
Length (English) 12 in.