Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Frit for Omnifit™ Column Endpiece

Catalog No. 111005083 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Columns - 10 mm Bore
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-100-5083 Columns - 10 mm Bore
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5083 Supplier MilliporeSigma™ Supelco™ Supplier No. 56102
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Frit's pore diameter and surface structures should be optimized to avoid clogging and abrasion of chromatographic particles.

  • End pieces are equipped with porous frits
  • Frits can be made of sintered metal, glass and plastic
  • Designed to be easily exchangeable
  • Can also be used to keep packing material in place
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Columns - 10 mm Bore
Material Polyethylene
Packaging Quantity 20 Pk.
Type Frit
Pore Size 25 μm

Omnifit is a registered trademark of Diba Industries

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.