Learn More
Novus Biologicals™ Frizzled-4 Overexpression Lysate

Description
Specifications
Specifications
| Cell Line | The lysate was created in HEK293T cells, using plasmid ID RC217286 and based on accession number NM_12193. The protein contains a C-terminal DDK tag. |
| For Use With (Application) | Western Blot |
| Quantity | 0.1 mg |
| Storage Buffer | RIPA buffer |
| Purity | Protein |
| Test Specificity | Homo sapiens frizzled homolog 4 (Drosophila) (FZD4), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Research Category | GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway |
| Protein State | Native |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.