Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frizzled 5/8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985020
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5020 Supplier Bioss Supplier No. BS2930R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Members of the ′frizzled′ gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD5 protein is believed to be the receptor for the Wnt5A ligand.

Specifications

Antigen Frizzled 5/8
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FZD5
Gene Accession No. Q13467
Gene Alias C2orf31; DKFZp434E2135; frizzled 5, seven transmembrane spanning receptor; frizzled class receptor 5; frizzled family receptor 5; Frizzled-5; Fz5; Fz-5; Fzd5; Fzd-5; FzE5; HFZ5; MGC129692; seven-transmembrane receptor frizzled-5; Wnt receptor
Gene Symbols FZD5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Frizzled 5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7855
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.