Learn More
Description
Includes
Micro Centrifuge only. Rotor is not included.
- Easy-to-Use Control Panel that Supports Simple Operation: The intuitive control panel with rotary knob puts the centrifuge’s simple operations at your fingertips, including the ability to set and recall speed
- Sophisticated Technology that Drives High-Performance: A powerful brushless induction motor, 10 acceleration and deceleration settings, and memory for up to 99 commonly-used parameters are a few of Frontier’s features that propel high-performance
- Safety Features that Protect Against Injury: Among Frontier’s safety attributes are a motorized lid locking system, and a rotor imbalance sensor that stops the centrifuge if the rotors become imbalanced or sudden shock occurs, protecting the operator from the high-speed spinning rotors
- Rotor Sizes that Provide Flexibility for Varied Centrifugation Needs: Compatible with seven different rotors, more than any centrifuge in its class, the highly-versatile Frontier can be used for various applications of different volumes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 12 x 5 or 4 x 8 PCR Strips |
| Max. Speed | 15,200 rpm |
| Max. RCF | 21,953 x g |
| Electrical Requirements | 120 V |
| Refrigerated | Yes |
| Frequency | 50/60 Hz |
| Voltage | 120/230 V |
| Rotor | Rotor is not included; sold separately. |
| Product Type | Centrifuge |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
