Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fructose, Granular, FCC, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602858 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-858 12 kg Poly Bottle
18-602-856 2.5 kg Poly Bottle
18-602-855 6 x 500 g Poly Bottle
18-602-857 4 x 2.5 kg Poly Bottle
18-602-854 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-858 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F109212KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F1092, 57-48-7, C6H12O6

Spectrum™ Chemical Fructose, Granular, FCC is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used commercially as a sweetener in food and beverages due to its high sweetness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO

Specifications

CAS Max % 100%
Ignition Residue 0.5%
Loss on Drying 0.5%
Packaging Poly Bottle
Quantity 12 kg
Percent Purity 98 to 102%
Grade FCC
Physical Form Crystals or Flakes
Chemical Name or Material Fructose, Granular
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.