Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 8 to 10 mL Blood Sample Tubes; 1 Drawer, holds 60 tubes
Supplier:  Cole-Parmer 0440175
Encompass_Preferred
These upright freezer drawer racks are specially designed for storing blood sample tubes. They are crafted of corrosion-resistent stainless steel for durablity.
Catalog No. 50-196-0519
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.