Learn More
Cole-Parmer Argos Technologies PolarSafe Vertical/Chest Freezer Rack for Microtube Boxes with 2 7/16" Max Height, 8 box capacity
Supplier:  Cole-Parmer 0440222
Encompass_Preferred
These upright freezer racks come with a locking rod to securely hold boxes in place. Boxes can be accessed from either side of the rack, which has handles on both sides. They are crafted of corrosion-resist stainless steel for long-term use.
Catalog No. 50-196-0774
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.