Learn More
Andwin Scientific FRZR BOX DIVIDER 49 HOLE 12/PK
SDP
Supplier:  Andwin Scientific 6161585
Encompass_Preferred
The Freezer Box Divider 49 Hole is used to create partitions in the cardboard freezer box. This product can be used to hold up to 49 cryogenic vials stored in liquid nitrogen or mechanical freezers. It is sold in a 12-pack.
Catalog No. 50-727-640
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.