Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FSD1L/CCDC10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982714
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2714 Supplier Bioss Supplier No. BS13221R
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FSD1L is a protein coding gene.

Specifications

Antigen FSD1L/CCDC10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FSD1L
Gene Accession No. Q9BXM9
Gene Alias A230072O16Rik; BOS_9310; CCDC10; coiled-coil domain containing 10; Coiled-coil domain-containing protein 10; CSDUFD1; cystatin and DUF19 domain containing 1; fibronectin type III and SPRY domain containing 1 like; fibronectin type III and SPRY domain containing 1-like; FSD1 C-terminal like; FSD1 N-terminal like; FSD1 N-terminal-like protein; FSD1CL; FSD1L; FSD1-like protein; FSD1NL; MIR1; OTTMUSG00000007002; RGD1564809; sb:eu990
Gene Symbols FSD1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FSD1L/CCDC10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83856
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.