Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FSH Native Protein
SDP

Catalog No. NB19952550U Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB19952550U 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB19952550U Supplier R&D Systems™ Supplier No. NBP19952550UG

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results.

  • Native bioactive protein containing 165 amino acids for Human FSH
  • The FSH Native Protein is derived from Urine of post-menopausal women.
  • This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Specifications

Concentration Lyoph
For Use With (Application) Bioactivity, SDS-PAGE
Molecular Weight (g/mol) 35.5 kDa
Purification Method >98%, by SDS-PAGE
Quantity 50 μg
Source Urine of post-menopausal women
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 ng/μg
Reconstitution Reconstitute in dH2O to a concentration of 0.1 - 1.0 mg/mL. The solution can then be diluted into other aqueous buffers and stored at 4°C for 1 week or -20°C for long-term storage.
Buffer Lyophilized.
Product Line Novus
Purity or Quality Grade >98%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.