Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ FTBMT
GREENER_CHOICESDP

Catalog No. 67845
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-845 5 mg
67-842-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-845 Supplier Tocris Bioscience™ Supplier No. 6784/5

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective GPR52 agonist; orally bioavailable and BBB permeable

FTBMT (also known as TP 024) is a potent and selective GPR52 agonist (EC50 = 75 nM, Emax = 122%), which is selective for GPR52 over a panel of 98 targets including D1, D2, AMPA and NMDA. FTBMT suppresses methamphetamine-induced hyperlocomotion in mice, and inhibits MK-801 -induced hyperactivity (model for acute psychosis), without causing catalepsy in mice. This compound is orally bioavailable and brain penetrant.

Specifications

Chemical Name or Material 4-[3-[[3-Fluoro-5-(trifluoromethyl)phenyl]methyl]-5-methyl-1H-1,2,4-triazol-1-yl]-2-methylbenzamide
CAS 1358575-02-6
Quantity 5 mg
Molecular Formula C19H16F4N4O
Formula Weight 392.35
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.