Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FTSI Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144458 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144458 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144458 Supplier Invitrogen™ Supplier No. PA5144458
  • $402.00
    $372.65 / Each of 1

    Save $29.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

FTSI Polyclonal Antibody for Western Blot, ELISA

Essential cell division protein that catalyzes cross-linking of the peptidoglycan cell wall at the division septum (PubMed:1103132, PubMed:6450748, PubMed:9614966, PubMed:3531167, PubMed:7030331). Required for localization of FtsN (PubMed:9282742).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSI
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene ftsI
Gene Accession No. P0AD68
Gene Alias pbpB
Gene Symbols ftsI
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) FTSI protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 75202099
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.