Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FTSJ2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733888 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3888 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3888 Supplier Proteintech Group Inc Supplier No. 225561AP150UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Putative ribosomal RNA methyltransferase 2 (FTSJ2), also named as FJH1 or Protein FTSJ homolog 2 is a 246 amino acid protein, which belongs to the methyl-transferase superfamily. FTSJ2 localizes in the nucleus and is widely expressed in in muscle, placenta, and heart. FTSJ2 functions as a nucleolar RNA methyl-transferase involved in eukaryotic RNA processing and modification.

Specifications

Antigen FTSJ2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MRM2
Gene Accession No. Q9CPY0, Q9UI43
Gene Alias FJH1, FtsJ homolog 2 (E. coli), FTSJ2, Protein ftsJ homolog 2
Gene Symbols MRM2
Host Species Rabbit
Immunogen FTSJ2 Fusion Protein Ag18410
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29960, 68017
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.