Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FtsZ (GTPase), Rabbit anti-Bacteria, Polyclonal, Millipore Sigma™

Catalog No. ABS220025UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS220025UL 25 μL
ABS2200100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS220025UL Supplier MilliporeSigma Supplier No. ABS220025UL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Anti-FtsZ (GTPase), Cat. No. ABS2200, is a rabbit polyclonal antibody that detects Cell division protein FtsZ and has been tested for use in Immunofluorescence and Western Blotting.

Cell division protein FtsZ (UniProt: P17865; also known as Cell Division FtsZ GTPase, FtsZ) is encoded by the FtsZ (also known as BSU15290) gene (Gene ID: 935971) in Bacillus subtilis. FtsZ is homodimeric prokaryotic homologue of tubulin with GTPase activity. It is found in nearly all bacteria. It polymerizes in the presence of GTP to form higher-ordered polymers. It contains three GTP-binding sites that are located in the N-terminal region. The less conserved C-terminal domain contains residues that are important for GTP hydrolysis. FtsZ is shown to be essential for bacterial cell division and in association with FtzA (FtsAZ) it forms a contractile ring structure (Z ring) at the site of cell division. The regulation of the ring assembly controls the timing and the location of cell division. One of the major functions of FtsZ is to recruit other cell division proteins to the septum to produce a new cell wall between the dividing cells. The FtsZ treadmilling rate controls the rate of peptidoglycan synthesis and cell division. During bacterial cell division FtsZ co-localizes with SpoIIE, a dual function protein that contributes to the switch form medial to polar cell division during sporulation. The anti-FtsZ blot has also been used as control for sample loading. (Ref.: Bisson-Filho, AW., et al., (2017). Science 355(6326); 739-743; Feucht, A., and Jeffery Errington, J (2005). Microbiology 151(6); 2053-2064; Lucet, I., et al. (2000). EMBO J. 19(7); 1467-1475). Lytvynenko, I., et al. (2019). Cell. 178(1); 76-90)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FtsZ (GTPase)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected FtsZ (GTPase) in Western Blotting applications (Lucet, I., et. al. (2000). EMBO J. 19(7):1467-75; Bisson-Filho, A.W., et. al. (2017). Science. 355(6326):739-743).Immunofluorescence Analysis: A representative lot detected FtsZ (GTPase) in Immunofluorescence applications (Daniel, R.A., et. al. (2000). Mol Microbiol. 35(2):299-311).
Formulation Rabbit polyclonal antiserum with 0.05% sodium azide.
Gene Alias Cell division protein FtsZ;Cell Division FtsZ GTPase
Gene Symbols FtsZ;BSU15290
Host Species Rabbit
Immunogen Full length purified FtsZ from Bacillus subtilis.
Molecular Weight of Antigen ∽44 kDa observed; 40.40 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Unpurified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_389412
Test Specificity This rabbit polyclonal antibody detects Cell Division Protein FTsZ in Bacillus subtilis.
Target Species Bacteria
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.