Learn More
AdipoGen FTY720 . hydrochloride (1 mg)
SDP
Supplier:  AdipoGen AGCR13587M001
Encompass_Preferred
FTY720 . hydrochloride (1 mg), AdipoGen Life Sciences. Chemical. CAS: 162359-56-0. Formula: C19H33NO2 . HCl. MW: 307.4 . 36.5. Synthetic analog of ISP-1, isolated from Isaria sinclairii. Cell permeable immunosuppressor displaying lymphocyte sequestration properties and used for treatment in multiple sclerosis. Potent sphingosine 1-phosphate (S1P) receptor (S1P1, S1P3, S1P4, and S1P5) agonist when phosphorylated by sphingosine kinase. Sphingosine transporter Abcb1 and leukotriene C4 transporter Abcc1 activity enhancer. Cytosolic phospholipase A2 inhibitor, independent of sphingosine 1-phosphate receptors. Autophagy inducer. Apoptosis inducer.
Catalog No. 50-149-0100
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.