Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUBP1 Recombinant Rabbit Monoclonal Antibody (1G6X5)
GREENER_CHOICE

Catalog No. PIMA542907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542907 Supplier Invitrogen™ Supplier No. MA542907
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: YQQQAQPPPA APAGAPTTTQ TNGQGDQQNP APAGQVDYTK AWEEYYKKMG QAVPAPTGAP PGGQPDYSAA WAEYYRQQAA YYAQTSPQGM PQHPPAPQGQ.

This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1G6X5
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FUBP1
Gene Accession No. Q32PX7, Q91WJ8, Q96AE4
Gene Alias 9530027K12Rik; D3Ertd330e; DNA helicase V; EMBL:AAI07943.1, ECO:0000312; EMBL:AAI07943.1}; far upstream element (FUSE) binding protein 1; far upstream element (FUSE) binding protein 4; far upstream element binding protein 1; far upstream element-binding protein; far upstream element-binding protein 1; far upstream element-binding protein 1 {ECO:0000250; FBP; FBP {ECO:0000250; Fubp; Fubp1; fubp1 {ECO:0000312; Fubp4; FUSE binding protein 1; FUSE-binding protein 1; FUSE-binding protein 1 {ECO:0000250; hDH V; RGD:1591892}; UniProtKB:Q96AE4, ECO:0000312; UniProtKB:Q96AE4}
Gene Symbols FUBP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 545-644 of human FUBP1 (Q96AE4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51886, 654496, 8880
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.