Learn More
Fuchsin-Carbol, Ziehl-Neelsen Formulation, for Staining Acid Fast Organisms, Ricca Chemical
Shop All Ricca Chemical Company Products

Description
- Ziehl-Neelsen. Requires special barrier pack at additional charge for shipment via UPS Air
- Glass/Poisonous Liquid
Specifications
Specifications
| Chemical Name or Material | Fuchsin-Carbol |
| Name Note | for Staining Acid Fast Organisms |
| CAS | 7732-18-5, 64-17-5, 108-95-2, 67-56-1, 569-61-9 |
| CAS Min % | 87.0, 6.95, 3.91, 0.26 |
| CAS Max % | 90.0, 7.23, 4.07, 0.27 |
| Grade | Laboratory |
| Packaging | Amber Glass Bottle |
| Color | Dark Red |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.