Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fuel Oil Degradation Mix, Restek™
SDP

Catalog No. 06801369 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-801-369 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-801-369 Supplier Restek Corporation Supplier No. 31240
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

  • Solutions are prepared from single-source (one-refinery) product
  • Weathered materials indicate the percent weight loss from the original material
  • Subsurface degradation of fuel oil spills can be estimated by examining ratios of C17/pristane and C18/phytane

Specifications

Packaging Ampule
CAS Min % 99.2, 0.200000, 0.200000, 0.200000, 0.200000
CAS 629-78-7, 638-36-8, 1921-70-6, 593-45-3
Odor Strong
Vapor Density 2.93 (Air = 1)
Specific Gravity 1.3254 - 1.3258 g/cm3 at 20°C
Concentration 2,000 μg/mL each in Methylene Chloride
UN Number UN1593
Color Colorless
Melting Point 96.7°C
Solubility Moderately soluble
Type Multi-component Standard
Quantity 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.