Learn More
TRANE Technologies FARRAR Lab Grade Upright ULT Freezer -86°C (FUF126)
Supplier:  TRANE Technologies 5818126FAR115
Encompass_Preferred
FARRAR’s Ultra-Low Temperature Freezer is SNAP/EU F-gas-compliant, offering low energy consumption with a low global warming potential (GWP) natural hydrocarbon refrigerant (R290/R170). With a temperature range of -50°C to -86°C with <+/-5°C uniformity, our ULT freezer features a 26ft3 (737L) capacity.An integrated Smart Access Panel features integrated, exportable temperature logs, a PIN controlled electronic door lock, and offers maximum protection for your samples with standard 5 inner doors. FARRAR Ultra-Low Temperature Freezers have undergone detailed Accelerated Life Testing (ALT) to deliver long service life in a busy lab environment.Specifications for model FUF126* Capacity - 26 cu ft (736 L)* Shelves - 4 adjustable shelves and variety of rack systems available* Exterior Dimensions - 40.2 x 78.2 x 35.1 in* Electrical - dual voltage: 115V 60Hz or 220-240V 50/60Hz
Catalog No. NC3257234
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.