Learn More
Thomas Scientific 500mL Autofil 0.2æm High Flow PES Bottle Top Filter, Full Assembly
SDP
Supplier:  Thomas Scientific 1198H42
Encompass_Preferred
Foxx Life Sciences’ Autofil Bottle-Top Filtration Systems are disposable, sterile, vacuum operated systems for the sterile preparation of buffers, tissue culture media, microbiological media, and other biological fluids. Systems are available with asymmetric 0.22 µm (sterilization grade) and 0.45 µm (clarification grade) PES membranes. The complete system is composed of a polystyrene filter funnel and receiver bottle, a SECUREgrasp cap, and a vacuum hose adapter. Systems with 15 mL and 50 mL polypropylene centrifuge tubes are also available. Filtration funnels are compatible with the patented Autofil pedestal, a cradle ring for use with existing lab stands, and vacuum hose adapters. All components are USP Class VI, FDA grade, and are conveniently sold gamma-sterilized. Quick-Connect pedestal offers one-time vacuum connection and one-handed operation Conveniently sold pre-sterilized Autofil is available as a complete system or as a bottle-top filter
Catalog No. 50-145-9573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.