Learn More
PIP™ Full Frame Safety Glasses
Catalog No. 19153414 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-153-414 Clear
19-153-415 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-153-414 Supplier PIP™ Supplier No. 250500520
  • $22.04
    $13.65 / Each
    Save $8.39
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,587.02
    $982.65 / Case of 72 EA
    Save $604.37
    38% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
PIP™ Full Frame Safety Glasses feature interchangeable temples and headband (included) to provide the proper protection for the job at hand.
  • Flame resistant soft foam padding forms a seal to keep out dust and airborne particles
  • Comes with both temples and elastic headband strap
  • Temples and strap snap in or out for easy conversion from a safety glass to a dust goggle and back
  • Bayonet nylon temples with rubber tips for a secure fit
  • Molded nose bridge design to fit the vast majority of users
  • FogLess™ 3Sixty™ enhanced hydrophobic action resists lens fogging and provides clear vision in extreme working conditions
  • 8 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Dust Goggles
Frame Color Black
Includes Foam padding; Headband
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Type Sealed with Foam
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Reproductive Harm