Learn More
Mopec Full-Length Shelf for Dissection Tables
SDP
Catalog No. 21403490 Shop All Mopec Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
21-403-490 Stainless Steel
1 options
Catalog No. 21-403-490 Supplier Mopec Supplier No. HO024
Add to Cart
Add to Cart
A specialized pathology grossing workstation accessory designed to improve efficiency and safety during specimen handling.
  • Construction:
    • Built with durable, lab-grade materials, it integrates seamlessly into Mopec grossing stations and spans the full length of the underside of the dissection table, providing organized storage and support for tools and workflow
  • Material:
    • 304 stainless steel construction with a polished No. 4 finish
  • Dimensions:
    • 82.4 (L) x 23.3 (W) x 1 (H) in.
    • Usable area 82.2 (L) x 19.6 (W) in.
  • Compatibility:
    • HA100
    • HA210
    • HA400
    • HB1400
    • HB1410
    • HB1500
    • HB1510
Specifications
Product Type Shelf
Autoclavable Not Autoclavable
Color Silver
Material Stainless Steel
Disposable Reusable
Sterility Non-sterile