Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Full-Size Disarticulated Skeleton Model
SCI_ED

Catalog No. S81003 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81003 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81003 Supplier Walter Products™ Supplier No. B10220
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

All 206 bones of the skeletal system represented. All bones are cast individually with anatomical detail.

  • High quality, life-size disarticulated skeleton made exclusively from high quality medical PVC
  • One hand and foot are wired, while the opposite hand and foot are detached to allow further study of individual bones
  • 3-part skull with removable skull cap and spring-mounted mandible
  • Color poster of the skeletal system included
  • Perfect for students and teachers

Specifications

For Use With (Application) Education
Includes Bones & instructional manual, Color poster of the skeletal system
Material PVC
Quantity 1
Weight (Metric) 7.35 kg
Product Type Model, Skeleton, Disarticulated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.