Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Full-Size Flexible Skeleton Model
SCI_ED

Catalog No. S81001 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S81001 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81001 Supplier Walter Products™ Supplier No. B10229
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This full-size human skeleton has a flexible spine that can bend to mimic natural human movement

  • High quality, durable life-size model made from high quality medical PVC
  • All movable parts are joined by connectors, and utilized in such a way to allow movement around the joint
  • Hands and feet are wire mounted to demonstrate their natural position
  • Both the arms and legs are removable for further study
  • The removable skull features a movable jaw, removable skullcap, suture lines and a full dentition
  • Bones are cast individually from naturally colored plastic with anatomical detail
  • Comes pelvic mounted on a roller base and supplied with a diagram
  • Key structures are numbered and sutures have been highlighted
  • Corresponding key card included

Specifications

For Use With (Application) Education
Height (Metric) 170 cm
Includes Skeleton & roller stand
Material PVC
Quantity 1
Size Life-Size
Weight (Metric) 11.79 kg
Product Type Model, Skeleton, Flexible
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.