Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Full-Size Hanging Skeleton with Muscles
SCI_ED

Catalog No. S80966 Shop All Walter Products Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S80966 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80966 Supplier Walter Products™ Supplier No. B10200H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Life-size human skeleton made of high quality medical PVC

  • Life-size human skeleton made of high quality medical PVC
  • Muscle origins (red) and insertions (blue) are accurately painted and numbered
  • Skull has movable jaw, removable skull cap, suture lines and a full dentition
  • Both arms and legs can be removed for closer study
  • Available pelvic mounted or hanging on a roller stand

Specifications

Color Off-white
For Use With (Application) Education
Height (Metric) 170 cm
Includes Comes on a roller base and supplied with diagram
Material PVC
Quantity 1
Size Life-size
Weight (Metric) 12 kg
Product Type Model, Human Skeleton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.