Learn More
Fullerene, buckytube/nanotube, single walled, Thermo Scientific Chemicals
Description
Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Quantity | 1 g |
UN Number | UN1325 |
Solubility Information | It is miscible,can be diluted with water. Soluble in benzene,toluene. |
Physical Form | Powder |
Chemical Name or Material | Fullerene, Buckytube/nanotube, single walled |
Safety and Handling
Hazard Category | H228-H315-H319-H335 |
Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.