Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fumaric Acid, NF, 99.5-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602872 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-872 12 kg Poly Bottle
18-602-874 100 g Poly Bottle
18-602-870 500 g Poly Bottle
18-602-871 2.5 kg Poly Bottle
18-602-873 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-872 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F109612KG
Add to Cart
Add to Cart
Request a Quote

F1096, 110-17-8, C4H4O4

Spectrum™ Chemical Fumaric Acid, NF is used an effervescent agent in tablet formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

CAS Max % 100%
Boiling Point 290°C
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 12 kg
Percent Purity 99.5 to 100.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Fumaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.