Learn More
FUMEX INC FUMEX FA100 REPLACEMENT FILTER
SDP
Supplier:  FUMEX INC FA100
Encompass
The 12” x 12” x 5.75” Fumex FA100 Replacement Filter incorporates high performance HEPA media composed of borosilicate microfibers with an acrylic resin binder. This glass microfiber media is tested and certified at the manufacturer as meeting HEPA media standards with a DOP maximum smoke penetration of 99.97% efficient at 0.3 µm @ 10.5 ft/minute. The media is tested to NSF standards ASTM method F778-82 for permeability and the ASTM method D-2986 for DOP smoke penetration.
Catalog No. NC1729459
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.